Fixed-angle titanium rotor package (26° angle), MLA-80
Leverancier: Beckman Coulter
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
367096
521-4041EA 17160 EUR
521-4041
Fixed-angle titanium rotor package (26° angle), MLA-80
Centrifuge Rotoren
Major applications: Large volume differential sedimentation (pelleting) of subcellular organelles and viruses.