Heptafluorobutyric acid, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid

M-Clarity™ quality level = MQ100

52411-5ML-F 52411-25ML-F
52411-5ML-FEA 50.6 EUR
52411-5ML-F 52411-25ML-F
Heptafluorobutyric acid, Supelco®
Heptafluorobutyric acid
Formule: C₄HF₇O₂
Kookpunt: 121 °C (1013 hPa)
Dichtheid: 1,764 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004171
CAS nummer: 375-22-4
EINECS: 206-786-3
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=501afc7dcddf2c57d0707b9e9b3e78454e64b58b&vid=ff9f462ce2d2fc1c55f667e4e1dd221218758ae7&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR