Axygen® Microcentrifuge Screw Caps
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SCO-B SCO-BR SCO-C SCO-G SCO-LP-B SCO-LP-BR SCO-LP-O SCO-LP-Y SCO-R SCO-Y SCO-V-S SCO-R-S
525-0814EA 108 EUR
525-0814 525-0815 525-0333 525-0819 525-0823 525-0824 525-0829 525-0835 525-0840 525-0846 525-0843 525-0841
Axygen® Microcentrifuge Screw Caps
Sluitingen Reageerbuissluitingen
These caps are compatible with all Axygen Scientific screw top microcentrifuge tubes, including both skirted and nonskirted variations of the 0,5 ml, 1,5 ml, and 2 ml microcentrifuge tubes.
  • Available looped and non-looped
The O-ring closure is formulated and manufactured from a special compound resistant to chemicals such as anhydrous ethyl alcohol, purified ether, hexane, acetone, chloroform, acetonitrile, 40% trifluoroacetic acid solution, and other chemicals used in most laboratories. The caps have extra long lips to help protect samples and ribsMore Product Information