Safe-Lock Tubes, 2,0 ml - Microtubes
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0030120.094 0030120.205 0030120.213 0030120.221 0030120.230 0030120.248 0030121.597 0030121.686 0030123.344 0030123.620
211-2120EA 71 EUR
211-2120 211-2121 211-2122 211-2123 211-2124 211-2125 211-2160 211-2129 211-2165 525-1043
Safe-Lock Tubes, 2,0 ml - Microtubes
Buisjes Microcentrifugebuisjes
Eppendorf® Safe-Lock 2,0 ml microtubes ensure secure sample preservation. With an innovative Safe-Lock mechanism, these microtubes prevent accidental openings, providing reliability in a 2,0 ml capacity. Ideal for various applications, they combine durability, ease of use, and sample protection.
  • Safe-Lock tube with a hinged lid prevents accidental opening during incubation and storage
  • g-Safe® technology, dependent on the Safe-Lock Tube variant, ensures exceptional centrifugation stability up to 30,000×g
  • g-Safe® technology prevents sample loss from tube
More Product Information