Eppendorf® DNA LoBind® Microcentrifuge Tubes
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0030108.078 0030108.035 0030108.051 0030108.310 0030122.208 0030122.232 0030122.348 0030108.400 0030108.418 0030108.426 0030108.523 0030122.275
525-0131EA 20.3 EUR
525-0131 525-0129 525-0130 525-0793 525-0985 525-0986 EPPE0030122.348 EPPE0030108.400 EPPE0030108.418 EPPE0030108.426 525-1602 EPPE0030122.275
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Buisjes Microcentrifugebuisjes
Unleash the power of precision in DNA research with Eppendorf® DNA LoBind® tubes. Engineered with proprietary LoBind® technology, these microcentrifuge tubes redefine sample handling, minimising the risk of loss and maximising DNA recovery. Ideal for low-concentration applications, including PCR, qPCR, and DNA sequencing, these tubes ensure accuracy, purity, and optimal yields.
  • Surface-coating-free, including substances like silicone, to reduce the risk of sample interference
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase,
More Product Information
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering Renewable Energy Certificate, ISO 14001