Centrifugeerbuisjes
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
103897 103797
525-2250EA 81.9 EUR
525-2250 525-2251
Centrifugeerbuisjes
Buisjes Centrifugebuisjes
PFA.
  • Translucent
  • Withstand forces up to 10000×g
  • Also for autosampler racks
Bestelinformatie: 525-2251 without screw cap, 525-2250 with calibrated ring-mark at 10 ml, thread GL25 and screw cap.
Opgelet: *The product(s) on this page are outside the scope of the EU Directive 2019/904