CentriVap® mobile systems
Leverancier: LABCONCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7812011 7812014
531-0169EA 18960 EUR
531-0169 531-0170
CentriVap® mobile systems
Verdampers Centrifugale verdampers
Combine the separate concentrator and cold trap components into one space-saving console and include casters for portability from laboratory to laboratory. A 12 to 17 mm rotor and solvent trap are also included. The interior can accommodate a vacuum pump - the only additional component necessary to begin processing.
  • Kamer uit aluminium met epoxycoating, en glazen deksel
  • Microprocessorgestuurd centrifugeren tot 1725 t/min en microprocessorgestuurde verwarming van 300 watt
  • Bedieningspaneel met aan/uit-knop, voorverwarmingsknop, optionele Heat
More Product Information