Verdampingskolven
Leverancier: BUCHI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
000431 000432 000433 000434 000435 000436 000437 020729 025323 025520
531-1811EA 91.5 EUR
531-1811 531-1812 531-1813 531-1814 531-1815 531-1816 531-1817 BUCH020729 BUCH025323 BUCH025520
Verdampingskolven
Kolven Kolven voor rotatieverdampers
Made of borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® R-210/215 rotary evaporators. Pear-shaped with NS 29.2/32 ground joint socket.