Drying flasks
Leverancier: BUCHI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
000452 000453 000454
531-1981EA 146 EUR
531-1981 531-1982 531-1983
Drying flasks
Kolven Kolven voor rotatieverdampers
Made of Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For rotary evaporators Rotavapor R-210/215. Pear shape with NS 29/32 neck. Glassware A, V, C.
Order Now
Specificaties
Environmentally Preferable