Nickel(II) chloride hexahydrate ≥99.95% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Nickel dichloride hexahydrate

53131.18 53131.A1 53131.30
53131.18EA 52.4 EUR
53131.18 53131.A1 53131.30
Nickel(II) chloride hexahydrate ≥99.95% (metals basis)
Nickel(II) chloride hexahydrate

Purity: 99.95% CAS No.: 7791-20-0 Molecular Formula: Cl2Ni.6H2O Formula Weight: 237.71 (129.60anhy) Physical Form: Crystalline Density: 1.92 Appearance: Green Odour: Odourless pH: 4.9 Melting Point: 140° -H2O Merck Reference: 14,6505 UN No.: 3288 MDL No.: MFCD00149809

Formule: NiCl₂·6H₂O
MW: 237.69 g/mol
Smeltpunt: 140 °C (-H₂O)
Dichtheid: 1.92
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149809
CAS nummer: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1 III
Merck Index: 14,6505

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR