Dimroth-condensatoren voor extractors, met glazen olijven
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05246004 05246006 05246007 05246008 05246005
533-2121EA 97.3 EUR
533-2121 533-2123 533-2127 537-0032 533-2122
Dimroth-condensatoren voor extractors, met glazen olijven
Condensors
DURAN® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Dimroth condensers for extractors, for condensing vapours. Water connection with two glass olives.