Sudan red 7B (fat red bluish, solvent red 19, fat red 7B), Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: N-Ethyl-1-((4-(phenyldiazenyl)phenyl)diazenyl)naphthalen-2-amine

M-Clarity™ quality level = MQ100

53373-25MG
53373-25MGEA 131 EUR
53373-25MG
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B), Supelco®
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Formule: C₂₄H₂₁N₅
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003904
CAS nummer: 6368-72-5
EINECS: 228-862-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba6b55793fcc211b343380e856603f2b2626a00f&vid=e4285187c2a237241625e9f6a5098938b3a06b8f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR