Ballotini solid soda glass balls, SiLibeads®
Leverancier: SIGMUND LINDNER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4507 5005 5009 5011 5012 5014 5016 5018 5005-20KG 4511-20KG 4504 4504-20KG 5003-20KG 4502-20KG 4507-20KG 5013 9609 5211-7 5210 4506 4505 4502 45015 5223-7 BL5016/10.0 5017 4511 BL5009/3.70-4. 5015 5216-7 5215-7 5214-7 5213-7 9611 45015-20KG 5212-7 5009-20KG 4505-20KG 9613 5011-20KG 5212-25KG 9623 5215-25KG
459-9131EA 0 EUR
459-9131 459-9132 459-9133 459-9134 459-9135 459-9136 459-9137 459-9138 201-0476 201-0474 201-0469 201-0470 201-0475 201-0468 201-0473 201-0477 201-0490 201-0481 201-0480 201-0472 201-0471 201-0467 201-0465 201-0489 LINRBL5016/10.0 201-0479 534-0002 LINRBL5009/3.70-4. 201-0478 201-0488 201-0487 201-0486 201-0485 201-0491 201-0466 201-0483 LINR5009-20KG 534-0001 534-0004 534-0003 201-0755 534-0005 LINR5215-25KG
Ballotini solid soda glass balls, SiLibeads®
Parels Glasparels
Ballotini solid glass balls are produced from high quality and pure soda-lime glass.
  • Main applications include:
  • Chemical plants (where large inert areas are required) and filters, example fractionating columns
  • Resin filler
  • Shot blasting
  • Burnishing (metal finishing)
  • Gas chromatography
The unique washing and polishing process does not involve the addition of hydrofluoric acid and thus gives the balls a pure shiny surface without contamination.