Freeze dryer, bench top, FreeZone®
Leverancier: LABCONCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
700201030 700202030 700401030 700402030 700801030 700802030 710201030 710202030 710401030 710402030 720401030 720402030
535-0393EA 10780 EUR
535-0393 535-0395 535-0397 535-0399 535-0387 535-0388 535-0401 535-0403 535-0405 535-0407 535-0416 535-0417
Freeze dryer, bench top, FreeZone®
Vriesdrogers
FreeZone® bench top freeze dryers are designed for lyophilising light to moderate sample loads. The compact design is ideal for laboratories with limited floor space.
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • Brushed stainless steel and glacier white, powder-coated steel exterior with blue accents
  • End-Zone™ end point detection option takes the guess work out of sample dryness
  • PTFE-coated collector coil and chamber for
More Product Information