VWR®, Screw Neck Vials ND8
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
548-0019AEA 284 EUR
548-0019A 548-0175A 548-2124A 548-0914A 548-1862A 548-0176A 548-0095A 548-0850A 548-2090A 548-0018A 548-0420A 548-0448A 548-0819A 548-0853A 548-0913A 548-0915A 548-1474A 548-1517A 548-1420A 548-1878A 548-0419A 548-1863A 548-2587A 548-2588A
VWR®, Screw Neck Vials ND8
flesjes
ND8 screw neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
  • Small opening
ND8 screw neck vials are constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear versions with a flat bottom to ensure compatibility with inserts. Associated screw caps, mostly available with assembled septa, are engineered to preventMore Product Information