TLC plates, special layers, glass backing, CHIRALPLATE
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
811057 811055 811056 811058 811059
552-0239EA 616 EUR
552-0239 552-0237 552-0238 552-0240 552-0241
TLC plates, special layers, glass backing, CHIRALPLATE
TLC-platen
These glass plates with a special layer are used for TLC enantiomer separations (e.g. amino acids, thiazolidine derivatives, dipeptides, lactones, α-hydroxycarboxylic acids).
  • Chiral separation based on ligand exchange
Base material: Reversed phase nano silica
Impregnation: Cu(II) ions and as chiral selector a proline derivative