TLC plates, silica gel 60, unmodified layers, aluminium sheets, ALUGRAM® Xtra SILG

Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

818232 818333 818331 818362 818329 818332 818360 818230.20 818261 818330.20
552-0326EA 85.8 EUR
552-0326 552-0329 552-0328 552-0331 552-1002 552-1005 552-1006 552-1000 552-1001 552-1004
TLC plates, silica gel 60, unmodified layers, aluminium sheets, ALUGRAM® Xtra SILG
TLC-platen
The highly polymeric binder used for these sheets is stable in almost all organic solvents and resistant towards aggressive visualisation reagents. It is also completely stable in purely aqueous eluents.

  • Precise colourisation results, even with 100% aqueous detection reagents
  • Easy and reliable cutting due to an optimised binder system, no flaking of silica

Silica 60: specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 to 17 μm.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR