Chromatography tubing, stainless steel
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
JR-T-97005 JR-T-97010 JR-T-97015 JR-T-97020 JR-T-97025 JR-T-97030 JR-T-97035 JR-T-97040 JR-T-97045 JR-T-97055 JR-T-97060 JR-T-97065 JR-T-97075 JR-T-97080 JR-T-97085 JR-T-97090 JR-T-97095 JR-T-97100 JR-T-97105 JR-T-97110
554-0796EA 25.1 EUR
554-0796 554-0797 554-0798 554-0799 554-1126 554-1127 554-1128 554-1129 554-1130 554-1131 554-1132 554-1133 554-1134 554-1135 554-1136 554-1137 JOURJR-T-97095 JOURJR-T-97100 JOURJR-T-97105 JOURJR-T-97110
Chromatography tubing, stainless steel
Slangen Chromatography Tubing
Stainless steel with PE caps and colour coded sleeves.
  • Ready to use
  • Cleaned
  • Soft annealed Ext.Ø for easy ferrule connections
  • For ¹/₁₆" zero dead volume connections
These tubes are cut to square, burr-free ends for zero dead volume connections. The pre-cut tubing is cut using an electrolytic cutting process and specially steam cleaned to remove both organic and inorganic contaminants. Plasticiser-free polyethylene caps are used to prevent contamination of the tubing.
Order Now
Specificaties
Environmentally Preferable