Stators, Rheodyne®
Leverancier: IDEX RHEODYNE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7010-040 7060-039 7060-065 7123-047 7125-067 7725-010 7725-026 9125-043 7900-146 7900-179 7900-183 7980-004
554-2061EA 938 EUR
554-2061 554-2086 554-2649 554-2090 RHEO7125-067 554-2150 554-2655 554-2189 554-2658 554-2659 554-2660 554-2664
Stators, Rheodyne®
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
Stators are available in 316 stainless steel, PEEK and proprietary materials. Rheodyne® stator materials have been researched and selected for their physical and mechanical strengths. Stators only need replacing if the ports or sealing surfaces become damaged.
DuraLife® stators are stainless steel stators with the wear surface coated to prolong valve life.
DuraLife® II stators are titanium base metal with coated wear surfaces.