Inline check valves, quick-stop luer, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
P-696
554-3155EA 248 EUR
554-3155
Inline check valves, quick-stop luer, Upchurch Scientific®
Ventielen Terugslagventielen
The Quick-Stop Luer check valve assembly is specifically designed to provide inline Luer connect/disconnect convenience while avoiding spills. It is made from biocompatible PEEK with gold-plated stainless steel spring.
  • Remains open when engaged
  • Designed to provide inline Luer connect/disconnect convenience without the mess and hazard of spills