Shodex Ohpak LB-800 Aqueous Size Exclusion Chromatography Columns for MALS Detectors
Leverancier: Resonac
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F6429202 F6429201 F6709434 F6429203 F6429204 F6429205 F6429206
554-5785EA 2522 EUR
554-5785 554-5781 554-5786 SHOWF6429203 SHOWF6429204 SHOWF6429205 SHOWF6429206
Shodex Ohpak LB-800 Aqueous Size Exclusion Chromatography Columns for MALS Detectors
Kolommen voor chromatografie
SEC columns for MALS detctors with ultra-low bleeding.
  • Same like OHpak SB-800 HQ series, but for light scattering detectors
  • Higher crosslinking for ultra-low bleeding for better chromatograms in particle-sensitive detectors
  • Polymer-based columns for aqueous SEC (GFC) analysis
  • Supports a wide range of molecular weight sample analysis
  • For modified proteins, PEGs, polysaccharides (hyaluronic acid, alginates), surfactants
  • The eluent can be replaced with DMF enabling the analysis of polar polymers
OHpak LB-800 are polymer-based SEC columns for theMore Product Information