HPLC columns, Hypersil GOLD™ C4
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25502-102130 25502-052130 25505-254630 25505-154630 25503-013001 25503-012101 25503-052130 25502-152130 25503-154630
555-5512EA 0 EUR
555-5512 554-4314 554-4316 555-5843 555-5519 555-5518 555-5527 555-5515 554-4315
HPLC columns, Hypersil GOLD™ C4
Kolommen voor chromatografie
These columns feature lower hydrophobicity than C18 or C8 and are recommended for very hydrophobic analytes.
  • Ideal for analysis of hydrophobic polypeptides and proteins
  • Highly symmetrical peak shapes
  • 1,9 µm particle size columns improve speed and efficiency