HPLC columns, GlycanPac™ AXH-1
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
082468 082469 082470 082472 082473 082474 082475 082476 082521
555-7539EA 1330 EUR
555-7539 555-7540 555-7541 555-7459 555-7363 555-7364 555-7365 555-7366 555-7371
HPLC columns, GlycanPac™ AXH-1
Kolommen voor chromatografie
These columns are high performance, silica-based HPLC columns for simultaneous separation of glycans by charge, size and polarity. They are designed for high-resolution and high-throughput analysis with unique selectivity for biologically important glycans, either labelled or native, by LC-fluorescence and LC-MS methods.
  • Useful for both high-resolution glycan profile characterisation and easy quantification of glycans based on charge
  • High chromatographic efficiency and excellent column stability
Particle type: high purity, spherical, porous silica