GC septa, TR-Blue
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
313B3233 313B3240
555-8258EA 0 EUR
555-8258 555-8259
GC septa, TR-Blue
Tussenschoten
Made of low-bleed silicone, these septa have excellent mechanical properties and are ideal for demanding GC-MS applications.
  • General purpose septa
  • Easy to penetrate
  • Maximum temperature 200 to 250 °C