Capillary columns, TraceGOLD™ TG-5HT
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
26095-0620 26095-0350 26095-0360 26095-0480 26095-1420 26095-1430 26095-0470 26095-1640
555-8736EA 1110 EUR
555-8736 555-8740 555-8739 555-8737 555-8734 555-8733 555-8738 555-8731
Capillary columns, TraceGOLD™ TG-5HT
Kolommen voor chromatografie
TraceGOLD™ TG-5HT columns are ideal for high temperature extended GC applications.
  • Low polarity
  • Lower bleed and better inertness than comparable high-temperature columns
  • Special design of fused silica tubing extends column lifetime by up to 40%