Capillary columns, TraceGOLD™ TG-POLAR
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
26082-0470 26082-0590 26082-5000 26082-5030 26082-5010 26082-5040 26082-5050 26082-5020
555-8994EA 879 EUR
555-8994 555-8992 555-8990 555-8987 555-8989 555-8986 555-8985 555-8988
Capillary columns, TraceGOLD™ TG-POLAR
Kolommen voor chromatografie
Specifically designed polymer and surface treatment overcome traditional problems with high-polarity columns.
  • Highly polar phase
  • Strong dipole moment and high selectivity for cis or trans compounds or compounds with conjugated double bonds
  • Equivalent to USP G8 and G48