Capillary columns, TRACE™ TR-WaxMS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
260X224P 260X296P 260X142P 260X154P
555-9089EA 0 EUR
555-9089 555-9087 555-9095 555-9092
Capillary columns, TRACE™ TR-WaxMS
Kolommen voor chromatografie
TRACE™ TR-WaxMS GC columns are ideal for high-polarity separations as part of GC/MS analysis.
  • Polar phase, polyethylene glycol
  • Proprietary bonding method for expanded operating temperatures
  • Extremely low bleed for improved sensitivity and library matches
  • High stability with oxygen and water
Order Now
Specificaties
Environmentally Preferable