Inlet base seals
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
290GA083 290GA084 290GA091 290GA092 290GA094 290GA122 290GA082 290GA081
555-9130EA 92.7 EUR
555-9130 555-9223 555-9222 555-9221 555-9219 555-9215 555-9037 555-9038
Inlet base seals
Afdichtingen Mechanische afdichtingen
Ideal for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenol.
Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports.