Capillary columns, TraceGOLD™ TG-1MS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
26099-2130 26099-2230 26099-2240 26099-2250 26099-2350 26099-2840 26099-2860 26099-2960 26099-2970 26099-2980 26099-3080 26099-3090 26099-3340 26099-3360 26099-2360 26099-1310 26099-0200 26099-1420 26099-1300 26099-2850 26099-1430 26099-1540 26099-1550 26099-2110 26099-0360 26099-2760 26099-2940 26099-4840 26099-5820
555-9322EA 667 EUR
555-9322 555-9321 555-9320 555-9319 555-9318 555-9376 555-9374 555-9372 555-9371 555-9370 555-9368 555-9367 555-9366 555-9365 555-9349 555-9330 555-9335 555-9329 555-9331 555-9375 555-9327 555-9326 555-9325 555-9324 555-9334 555-9377 555-9373 555-9359 555-9356
Capillary columns, TraceGOLD™ TG-1MS
Kolommen voor chromatografie
These non polar TraceGOLD™ TG-1MS GC columns are ideal for optimal signal-to-noise ratio, sensitivity and MS integrity for a wide variety of applications. These low bleed, stable columns achieve consistent results.
  • Non polar, 100% dimethyl polysiloxane
  • Equivalent to USP G2
  • Exceptionally low bleed
These columns are recommended for hydrocarbons, GROs, drugs of abuse and PCBs.