Capillary columns, TRACE™ TR-FAME
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
260M154P 260M155P 260M096P 260M141P 260M142P 260M143P 260M147P 260M149P 260M166L
555-9479EA 1420 EUR
555-9479 555-9478 555-9488 555-9485 555-9484 555-9483 555-9482 555-9481 555-9477
Capillary columns, TRACE™ TR-FAME
Kolommen voor chromatografie
TRACE™ TR-FAME GC columns are ideal for separation of cis or trans isomers and fatty acid methyl esters (FAMEs) with various degrees of unsaturation.
  • Low bleed for MS, even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance
  • Polar phase, 70% cyanopropyl polysilphenylene-siloxane
  • Higher operating temperature than other columns
These columns are also suitable for any application that requires a high-polarity column for use with MS detection.