Zoutzuur 1 N, BAKER ANALYZED® Volumetric, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Hydrochloric acid solution, Muriatic acid

M.O.C. Category NI

5620.9010EA 154.01 EUR
5620.9010 5620.1000 5620-02 5620-07
Zoutzuur 1 N, BAKER ANALYZED® Volumetric, J.T.Baker®
Zoutzuur
Formule: HCl
MW: 36.46 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance (Clear, colorless liquid) Passes Test
Normality 0.995 - 1.005
Sulfate (SO₄) ≤ 1 ppm
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Heavy Metals ≤ 0.5 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR