2-Fenoxyethanol, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ100

56753-1ML-F 56753-5ML-F
56753-1ML-FEA 45.8 EUR
56753-1ML-F 56753-5ML-F
2-Fenoxyethanol, Supelco®
2-Fenoxyethanol
Formule: C₆H₅OCH₂CH₂OH
Kookpunt: 245 °C (1013 hPa)
Smeltpunt: 13 °C
Dichtheid: 1,1094 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002857
CAS nummer: 122-99-6
EINECS: 204-589-7
Merck Index: 12,07410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8718648391680628b2d3cf76f5cf26ffb77988ee&vid=3fd8b3273a938553dccb3f2ccd0836ca193f9c17&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR