Gas purifiers, GateKeeper® GPU 70 4R
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
GPUS70FIX04R00 GPUS70FHX04R00 GPUS70FOP04R00
568-0925EA 0 EUR
568-0925 568-0924 568-0926
Gas purifiers, GateKeeper® GPU 70 4R
Filters Gasfilters
GateKeeper® GPU gas purifiers were designed to improve purity consistency, lifetime and final purity. Leveraging its expertise in material science, Entegris has developed a new generation of purification products that deliver unsurpassed value and performance.
  • Purity control to ppt levels
  • Low pressure drop across the purifier
  • Regenerable
  • Long lifetime
  • Use ambient temperature purification – no heat or power required to operate
  • Insensitive to orientation (horizontal/vertical)
The model 70 is designed for peak flow rates up to 5 slpm.
Certificaten: CEMore Product Information