Piperidine ≥99.5%, Biotechnology Grade, Burdick & Jackson™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

571261-200ML 571261-450ML
571261-200MLEA 143 EUR
571261-200ML 571261-450ML
Piperidine ≥99.5%, Biotechnology Grade, Burdick & Jackson™
Piperidine
Formule: C₅H₁₁N
Kookpunt: 107 °C
Smeltpunt: –10 °C
Dichtheid: 0,86 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005979
CAS nummer: 110-89-4
EINECS: 203-813-0
UN: 2401
ADR: 8,I
Merck Index: 12,07621

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.50 %
Refractive index (n 20/D) 1.451 - 1.455
Water (Karl Fischer) Max. 0.3 %
Absorbance at 370 nm Max. 0.050
Absorbance at 290 nm Max. 1.000
Refractive Index n20/D 1.452 (lit.)
Vapor Density 3 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR