SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate) ≥97% (by HPLC), Millipore®

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ100

573114-100
573114-100EA 227 EUR
573114-100
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate) ≥97% (by HPLC), Millipore®
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)

Heterobifunctional reagent useful for enzyme labeling of antibodies and antibody fragments. Cross-links both a sulfhydryl group and a free amine group. Useful for sequential conjugation to minimize polymerization.
Reacts with primary amines and sulfhydryl groups.

Formule: C₁₆H₁₈N₂O₆
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00009634
CAS nummer: 64987-85-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR