J.T.Baker® BAKER INSTRA-ANALYZED®, Lead Standard Solution, 1000 mg/L Pb in 5% Nitric Acid (from Pb) Standard for ICP
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5765.0100EA 0 EUR
5765.0100
J.T.Baker® BAKER INSTRA-ANALYZED®, Lead Standard Solution, 1000 mg/L Pb in 5% Nitric Acid (from Pb) Standard for ICP
Standaarden Kationische standaarden Lood standaarden
Certificaten: Traceable to NIST.