Supelco® BIOshell™ HPLC Columns, 160 Å Peptide Phenyl-Hexyl Fused-Core®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
577523-U 577524-U 577525-U 577526-U 577527-U 577528-U 577529-U 577530-U 577531-U 577532-U 577533-U 577534-U 577535-U 577536-U 577537-U 577538-U 577539-U 577540-U 577541-U 577542-U 577543-U 577545-U 577546-U 577547-U 577548-U 577550-U 577549-U 577544-U
577523-UEA 673 EUR
577523-U 577524-U 577525-U 577526-U 577527-U 577528-U 577529-U 577530-U 577531-U 577532-U 577533-U 577534-U 577535-U 577536-U 577537-U 577538-U 577539-U 577540-U 577541-U 577542-U 577543-U 577545-U 577546-U 577547-U 577548-U 577550-U 577549-U 577544-U
Supelco® BIOshell™ HPLC Columns, 160 Å Peptide Phenyl-Hexyl Fused-Core®
Kolommen voor chromatografie
BIOshell™ 160 Å Peptide Phenyl-Hexyl is a highspeed, high-performance liquid chromatography column based on a 160 Å Fused-Core® particle design.
  • Higher resolutions and higher peak capacities of peptides at ~50% backpressures of sub-2 µm fully porous paticles (FPP)-packed columns
  • Lower backpressure allows for columns to be used in series to maximise resolutions and peak capacity of complex proteomic samples or tryptic digests
  • Compatible with all UHPLC and LC-MS instrumentation
The particles provides a thn porous shell of high-purity silica surroundingMore Product Information