Supelco® Ascentis® C18 HPLC Columns
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
581300-U 581301-U 581302-U 581304-U 581308-U 581320-U 581321-U 581322-U 581323-U 581324-U 581325-U 581326-U 581342-U 581343-U 581346-U 581347-U 581370-U 581372-U 581373-U 581374-U 581376-U 581377-U 581379-U
581300-UEA 586 EUR
581300-U 581301-U 581302-U 581304-U 581308-U 581320-U 581321-U 581322-U 581323-U 581324-U 581325-U 581326-U 581342-U 581343-U 581346-U 581347-U 581370-U 581372-U 581373-U 581374-U 581376-U 581377-U 581379-U
Supelco® Ascentis® C18 HPLC Columns
Kolommen voor chromatografie
Ascentis® C18 HPLC columns are optimised to the three terms of the resolution equation: efficiency, retention, and selectivity. The classical C18 column is suitable for any method that specifies a C18-type column.
  • High purity, type B silica for inertness, reproducibility, and stability
  • Modern bonding process optimises bonded phase coverage and maximises stability, while minimising bleed and unwanted secondary interactions
  • Compatible with LC-MS and all of today's sensitive instruments and methods
  • Scalable sensitivity from analytical to preparative
  • High
More Product Information