Lithiumchloride 99, granular, AR® ACS, Macron Fine Chemicals™

Leverancier: Avantor Performance Materials
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

5852-04EA 0 EUR
5852-04 5852-06
Lithiumchloride 99, granular, AR® ACS, Macron Fine Chemicals™
Lithium chloride

Granular.

Formule: LiCl
MW: 42.39 g/mol
Kookpunt: 1382
Smeltpunt: 614
Dichtheid: 2.07 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011078
CAS nummer: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 99 %
ACS - Assay (LiCl) ≥99 %
Solubility Passes Test
ACS - Titrable Base (meq/g) ≤0.008
ACS - Calcium (Ca) ≤0.01 %
ACS - Heavy Metals (as Pb) ≤0.002 %
ACS - Insoluble Matter ≤0.01 %
ACS - Iron (Fe) ≤0.001 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.20 %
ACS - Sulfate (SO₄) ≤0.01 %
ACS - Barium (Ba) ≤0.003 %
ACS - Loss on Drying at 105°C ≤1.0 %
Appearance (colorless, hygroscopic, crystalline granules with some black specks)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR