Tube tags, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M560-01
590-0219EA 48.6 EUR
590-0219
Tube tags, b.safe
Etiketten
Flexible tube tag to stick on capillaries and tubes with Ext.Ø 0,4 to 6,35 mm, assorted colours (5 colours per pack).
  • Markable with ball pens
  • Assorted colours
  • Made of high quality PP
Coloured tags to identify capillaries and tubes. Markable with ball pens (removable) and permanent markers, or self-adhesive labels.