Antistatic mats, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R630-01
590-0346EA 273 EUR
590-0346
Antistatic mats, b.safe
Vloermatten Permanente vloermatten
Mat made of static dissipative material with permanently mounted grounding cable including clip for earthing connection.
  • Suitable grounding accessories for b.safe products
  • Customised versions can be offered on request
For grounding vessels made of static dissipative materials or persons before handling with easy flammable liquids.
Order Now
Specificaties
Environmentally Preferable