99.0-101.0% (by titrimetric analysis), LiChropur™ suitable for HPLC, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

59678-250G
59678-250GEA 184 EUR
59678-250G
99.0-101.0% (by titrimetric analysis), LiChropur™ suitable for HPLC, Supelco®
di-Potassium hydrogen phosphate
Formule: K₂HPO₄
MDL Nummer: MFCD00011383
CAS nummer: 7758-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR