Tris(hydroxypropyl)phosphine ≥80% (by 31P-NMR), Millipore®

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: THPP, THP, Tris(3-hydroxypropyl)phosphine

Packaged under inert gas.

598250-1
598250-1EA 137 EUR
598250-1
Tris(hydroxypropyl)phosphine ≥80% (by 31P-NMR), Millipore®
Tris(hydroxypropyl)phosphine

A water-soluble, neutral, and odorless disulfide reducing agent that is more stable and effective than DTT. THP is comparable with TCEP as a reducing agent. Its reduction capacity against oxidised glutathione is 0,232 meq/g (TCEP has a reduction capacity of 0,182 meq/g). THP is also more resistant to air oxidation as compared to DTT.

Formule: C₉H₂₁O₃P
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00269835
CAS nummer: 4706-17-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Form Colorless viscous liquid
Purity ≥80% by P-NMR
Storage +2 °C to +8 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR