Potassium chloride 1.0 M KCl volumetric solution, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Potassiumchloride

60121-1L
60121-1LEA 149 EUR
60121-1L
Potassium chloride 1.0 M KCl volumetric solution, Fluka™
Potassium chloride
Formule: KCl
Dichtheid: 1 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011360
CAS nummer: 7447-40-7
EINECS: 231-211-8

Order Now

Specification Test Results

Factor 0.995 - 1.005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR