Potassium chloride ≥99.5% (by argentometric titration), Puriss. p.a.

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

60130-250G 60130-1KG 60130-5KG 60130-6X1KG
60130-250GEA 38 EUR
60130-250G 60130-1KG 60130-5KG HONC60130-6X1KG
Potassium chloride ≥99.5% (by argentometric titration), Puriss. p.a.
Potassium chloride
Formule: KCl
Kookpunt: 1,413 °C (1.013 hPa)
Smeltpunt: 772 °C
Dichtheid: 2 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011360
CAS nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

Assay (argentometric) 99.5 - 101.0 %
pH (5 %, 20°C) 5.5 - 8.0
Calcium (Ca) Max. 0.005 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Chromium (Cr) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0002 %
Magnesium (Mg) Max. 0.001 %
Manganese (Mn) Max. 0.0005 %
Sodium (Na) Max. 0.02 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Bromide (Br) Max. 0.05 %
Iodide (I) Max. 0.002 %
Phosphate (PO₄) Max. 0.0005 %
Sulfate (SO₄) Max. 0.003 %
Total N Max. 0.001 %
Colour of the solution complying
Turbidity of solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR