Electrolyte filling solution for Ag/AgCl reference electrodes, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
60137-250ML 60137-1L
60137-250MLEA 27.9 EUR
60137-250ML 60137-1L
Electrolyte filling solution for Ag/AgCl reference electrodes, Supelco®
Elektrolyten Vuloplossingen voor elektrodes
~3 M Potassium chloride solution saturated with silver chloride (~0,16 M AgCl).