Potassium ionophore III, Selectophore™, function tested ionophore, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 2-Dodecyl-2-methyl-1,3-propanediyl bis[N-[5′-nitro(benzo-15-crown-5)-4′-yl]carbamate], 2-Dodecyl-2-methylpropane-1,3-diyl bis((16-nitro-2,3,5,6,8,9,11,12-octahydrobenzo[b][1,4,7,10,13]pentaoxacyclopentadecin-15-yl)carbamate), BME 44

M-Clarity™ quality level = MQ100

60397-100MG 60397-25MG
60397-100MGEA 634 EUR
60397-100MG 60397-25MG
Potassium ionophore III, Selectophore™, function tested ionophore, Supelco®
Potassium ionophore III

Potassium ionophore III may be used to formulate the ion-selective poly(vinyl chloride) (PVC) membranes as a part of potentiometric sensors used in potentiometric electric tongue for food analysis.


Ionophore used in solvent polymeric membrane electrodes for the assay of K+, especially in body fluids.


Verpakking: Bottomless glass bottle. Contents are inside inserted fused cone.

Formule: C₄₆H₇₀N₄O₁₈
MW: 967.06 g/mol
MDL Nummer: MFCD00145150
CAS nummer: 99348-39-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2079be3a93d4865f0273e1380049d3dbad9d350&vid=846c4d53285ff0ac742538540e52ac2c1b33c404&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR