Kiezelgel, Pore size: 60 Å 220-440 mesh, high purity for flash chromatography, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

60738-1KG 60738-5KG
60738-1KGEA 194 EUR
60738-1KG 60738-5KG
Kiezelgel, Pore size: 60 Å 220-440 mesh, high purity for flash chromatography, Supelco®
Kiezelgel
Formule: SiO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011232
CAS nummer: 112926-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba57e6ec4d0e34e8a8ad0a79a90dabae1ccb96da&vid=52422449495151fabda800a32121e9a8bce78ac7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR