VWR®, Microweegschuitjes
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
611-1355EA 232 EUR
611-1355 611-1356 611-1357 611-1358 611-1359 611-1360 611-1361
VWR®, Microweegschuitjes
Schuitjes Weegschuitjes
Aluminium.
These micro weighing boats are compatible with most commercially available analysers and microbalances, and are packed in easy to use plastic containers for storage and dispensing.