Pasteurpipetten
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2600145 2600155 2600119 2600135 2600111 2600151 2600120 2600190 2600171 2600195 2600181 2655111 2655120 2655151 2655181 2600131 2600216 2655171 2600160 2655135
612-1732EA 75.7 EUR
612-1732 612-1735 612-1740 612-1744 612-1746 612-1755 612-1762 612-1774 612-1776 612-5473 612-5472 612-5474 612-5475 RATI2655151 RATI2655181 RATI2600131 RATI2600216 RATI2655171 612-5471 RATI2655135
Pasteurpipetten
Pipetten Pasteur-/transferpipetten
PE, voor eenmalig gebruik.
  • Soft, elastic bulb
  • Unbreakable, reduced risk of injury compared to glass
  • Inert to biological fluids and most acids
  • Graduated pipettes with clearly visible embossed graduations
  • Sterile pipettes are gas-sterilised and individually packed
  • Suitable for food contact